gRNA Design Details
- Target DNA Sequence
- CGACTGGTTGTGCACGCGCA
- PAM Sequence
- CGG
- Target locus
-
Chr.2: 105242535 - 105242557 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
94.55/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- GPR45
- Entrez Gene ID
-
11250
- Gene Name
- G protein-coupled receptor 45
- Gene Aliases
-
PSP24, PSP24(ALPHA), PSP24A
- Chromosome Location
-
Chr.2: 105241743 - 105243467 on GRCh38
- UniGene ID
- Hs.590903
- Species
-
Human