gRNA Design Details
- Target DNA Sequence
- GGAGAATGTGGCGCTGAAGG
- PAM Sequence
- TGG
- Target locus
-
Chr.2: 120806822 - 120806844 on GRCm38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
84.85/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Ttbk2
- Entrez Gene ID
-
140810
- Gene Name
- tau tubulin kinase 2
- Gene Aliases
-
2610507N02Rik, AI326283, B930008N24Rik, TTK, Ttbk, Ttbk1, mKIAA0847
- Chromosome Location
-
Chr.2: 120732816 - 120850584 on GRCm38
- UniGene ID
- Mm.275698
- Species
-
Mouse