gRNA Design Details
- Target DNA Sequence
- GTAAAACCTCACAGCATCGC
- PAM Sequence
- TGG
- Target locus
-
Chr.9: 63616725 - 63616747 on GRCm38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
88.81/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Aagab
- Entrez Gene ID
-
66939
- Gene Name
- alpha- and gamma-adaptin binding protein
- Gene Aliases
-
2310007F21Rik, p34
- Chromosome Location
-
Chr.9: 63602629 - 63641897 on GRCm38
- UniGene ID
- Mm.34338
- Species
-
Mouse