gRNA Design Details
- Target DNA Sequence
- CTACAATGTGAACGACGAGA
- PAM Sequence
- AGG
- Target locus
-
Chr.15: 25364855 - 25364877 on GRCm38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
80.07/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Basp1
- Entrez Gene ID
-
70350
- Gene Name
- brain abundant, membrane attached signal protein 1
- Gene Aliases
-
2610024P12Rik, CAP-23, CAP23, Ckap3, NAP-22, NAP22
- Chromosome Location
-
Chr.15: 25363277 - 25413764 on GRCm38
- UniGene ID
- Mm.29586
- Species
-
Mouse