gRNA Design Details
- Target DNA Sequence
- ACCAATTCTCCACTCACCCA
- PAM Sequence
- CGG
- Target locus
-
Chr.18: 3511361 - 3511383 on GRCm38
- Strand
- Forward
- Application
- Gene Knockout
Score:
83.07/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Bambi
- Entrez Gene ID
-
68010
- Gene Name
- BMP and activin membrane-bound inhibitor
- Gene Aliases
-
2610003H06Rik
- Chromosome Location
-
Chr.18: 3507957 - 3516404 on GRCm38
- UniGene ID
- Mm.284863
- Species
-
Mouse