gRNA Design Details
- Target DNA Sequence
- AGCTGCCATCAGCGAGTCTG
- PAM Sequence
- TGG
- Target locus
-
Chr.9: 22380182 - 22380204 on GRCm38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
86.05/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Anln
- Entrez Gene ID
-
68743
- Gene Name
- anillin, actin binding protein
- Gene Aliases
-
1110037A17Rik, 2900037I21Rik, C78101, Scraps
- Chromosome Location
-
Chr.9: 22331214 - 22389696 on GRCm38
- UniGene ID
- Mm.282751
- Species
-
Mouse