gRNA Design Details
- Target DNA Sequence
- CTGCTACACGTCGTGTAGCC
- PAM Sequence
- AGG
- Target locus
-
Chr.14: 55576384 - 55576406 on GRCm38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
92.09/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Fitm1
- Entrez Gene ID
-
68680
- Gene Name
- fat storage-inducing transmembrane protein 1
- Gene Aliases
-
1110028A07Rik, AV010547, Cg10671, Fit1
- Chromosome Location
-
Chr.14: 55575674 - 55576952 on GRCm38
- UniGene ID
- Mm.36758
- Species
-
Mouse