gRNA Design Details
- Target DNA Sequence
- TTGCCAGCCCAGCTGTTGTG
- PAM Sequence
- AGG
- Target locus
-
Chr.11: 99597940 - 99597962 on GRCm38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
75.77/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Krtap9-3
- Entrez Gene ID
-
75586
- Gene Name
- keratin associated protein 9-3
- Gene Aliases
-
2310040M23Rik
- Chromosome Location
-
Chr.11: 99597348 - 99598106 on GRCm38
- UniGene ID
- Mm.75707
- Species
-
Mouse