gRNA Design Details

Target DNA Sequence
TTTGGGGGTGGCATTGGCTG
PAM Sequence
TGG
Target locus
Chr.11: 99597882 - 99597904 on GRCm38
Strand
Reverse
Application
Gene Knock-Out
Score: 70.47/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
Krtap9-3
Entrez Gene ID
75586
Gene Name
keratin associated protein 9-3
Gene Aliases
2310040M23Rik
Chromosome Location
Chr.11: 99597348 - 99598106 on GRCm38
UniGene ID
Mm.75707
Species
Mouse

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_029351.2 NP_083627.2 1