gRNA Design Details
- Target DNA Sequence
- CTCGGAGATACAATTTCAGC
- PAM Sequence
- AGG
- Target locus
-
Chr.5: 102875722 - 102875744 on GRCm38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
89.48/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Arhgap24
- Entrez Gene ID
-
231532
- Gene Name
- Rho GTPase activating protein 24
- Gene Aliases
-
0610025G21Rik
- Chromosome Location
-
Chr.5: 102207198 - 102897937 on GRCm38
- UniGene ID
- Mm.440191
- Species
-
Mouse