gRNA Design Details
- Target DNA Sequence
- CGAGGACGAAAAGCACAAAC
- PAM Sequence
- AGG
- Target locus
-
Chr.10: 79979729 - 79979751 on GRCm38
- Strand
- Forward
- Application
- Gene Knockout
Score:
83.96/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Tmem259
- Entrez Gene ID
-
216157
- Gene Name
- transmembrane protein 259
- Gene Aliases
-
AC087114.7gm2, ORF61, R32184_3, mFLJ00277
- Chromosome Location
-
Chr.10: 79977120 - 79984330 on GRCm38
- UniGene ID
- Mm.29442
- Species
-
Mouse