gRNA Design Details

Target DNA Sequence
ACGCGGACTTGTACACTTCC
PAM Sequence
CGG
Target locus
Chr.2: 109723665 - 109723687 on GRCm38
Strand
Forward
Application
Gene Knock-Out
Score: 87.95/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
Bdnf
Entrez Gene ID
12064
Gene Name
brain derived neurotrophic factor
Gene Aliases
-
Chromosome Location
Chr.2: 109674700 - 109727043 on GRCm38
UniGene ID
Mm.1442
Species
Mouse

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001048139.1 NP_001041604.1 2
NM_001048141.1 NP_001041606.1 2
NM_001048142.1 NP_001041607.1 2
NM_001285416.1 NP_001272345.1 2
NM_001285417.1 NP_001272346.1 2
NM_001285418.1 NP_001272347.1 2
NM_001285419.1 NP_001272348.1 2
NM_001285420.1 NP_001272349.1 2
NM_001285421.1 NP_001272350.1 2
NM_001285422.1 NP_001272351.1 1
NM_007540.4 NP_031566.4 2