gRNA Design Details

Target DNA Sequence
AGCTCGCTCTTCTGTTGCCA
PAM Sequence
AGG
Target locus
Chr.1: 91152255 - 91152277 on GRCm38
Strand
Reverse
Application
Gene Knock-Out
Score: 82.06/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
Rbm44
Entrez Gene ID
329207
Gene Name
RNA binding motif protein 44
Gene Aliases
Gm817
Chromosome Location
Chr.1: 91144973 - 91170799 on GRCm38
UniGene ID
Mm.295940
Species
Mouse

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001033408.4 NP_001028580.1 3