gRNA Design Details
- Target DNA Sequence
- AGCTCGCTCTTCTGTTGCCA
- PAM Sequence
- AGG
- Target locus
-
Chr.1: 91152255 - 91152277 on GRCm38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
82.06/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Rbm44
- Entrez Gene ID
-
329207
- Gene Name
- RNA binding motif protein 44
- Gene Aliases
-
Gm817
- Chromosome Location
-
Chr.1: 91144973 - 91170799 on GRCm38
- UniGene ID
- Mm.295940
- Species
-
Mouse