gRNA Design Details
- Target DNA Sequence
- AGGCGTTACTAAAGATGGCT
- PAM Sequence
- GGG
- Target locus
-
Chr.4: 12079944 - 12079966 on GRCm38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
86.48/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Tmem67
- Entrez Gene ID
-
329795
- Gene Name
- transmembrane protein 67
- Gene Aliases
-
5330408M12Rik, B230117O07, b2b1163.1Clo, b2b1291.1Clo
- Chromosome Location
-
Chr.4: 12039355 - 12089365 on GRCm38
- UniGene ID
- Mm.475125
- Species
-
Mouse