gRNA Design Details
- Target DNA Sequence
- AGCGCAGAACAGCTACCACG
- PAM Sequence
- AGG
- Target locus
-
Chr.5: 100086231 - 100086253 on GRCm38
- Strand
- Forward
- Application
- Gene Knockout
Score:
90.94/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Tmem150c
- Entrez Gene ID
-
231503
- Gene Name
- transmembrane protein 150C
- Gene Aliases
-
-
- Chromosome Location
-
Chr.5: 100077872 - 100165604 on GRCm38
- UniGene ID
- Mm.99790
- Species
-
Mouse