gRNA Design Details
- Target DNA Sequence
- CGATGAAGAAATCCACAACG
- PAM Sequence
- TGG
- Target locus
-
Chr.5: 100083721 - 100083743 on GRCm38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
85.96/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Tmem150c
- Entrez Gene ID
-
231503
- Gene Name
- transmembrane protein 150C
- Gene Aliases
-
-
- Chromosome Location
-
Chr.5: 100077872 - 100165604 on GRCm38
- UniGene ID
- Mm.99790
- Species
-
Mouse