gRNA Design Details
- Target DNA Sequence
- ACTCTGAGCATTGTACCGGG
- PAM Sequence
- TGG
- Target locus
-
Chr.1: 31196785 - 31196807 on GRCm38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
86.61/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Lgsn
- Entrez Gene ID
-
266744
- Gene Name
- lengsin, lens protein with glutamine synthetase domain
- Gene Aliases
-
Gluld1, Lgs, lengsin
- Chromosome Location
-
Chr.1: 31176435 - 31204725 on GRCm38
- UniGene ID
- Mm.389093
- Species
-
Mouse