gRNA Design Details
- Target DNA Sequence
- GACTTACAACAGGTTGGCTT
- PAM Sequence
- TGG
- Target locus
-
Chr.12: 102744711 - 102744733 on GRCm38
- Strand
- Forward
- Application
- Gene Knockout
Score:
87.08/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Tmem251
- Entrez Gene ID
-
320351
- Gene Name
- transmembrane protein 251
- Gene Aliases
-
D230037D09Rik
- Chromosome Location
-
Chr.12: 102743687 - 102745406 on GRCm38
- UniGene ID
- Mm.86546
- Species
-
Mouse