gRNA Design Details
- Target DNA Sequence
- GGGCATGGACTGTAATGGGG
- PAM Sequence
- AGG
- Target locus
-
Chr.7: 64695399 - 64695421 on GRCm38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
86.76/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Apba2
- Entrez Gene ID
-
11784
- Gene Name
- amyloid beta (A4) precursor protein-binding, family A, member 2
- Gene Aliases
-
X11-like, X11L, XllL, mXllL
- Chromosome Location
-
Chr.7: 64501706 - 64753878 on GRCm38
- UniGene ID
- Mm.4657
- Species
-
Mouse