gRNA Design Details
- Target DNA Sequence
- TAGTGAAGGGAGACAGCCGC
- PAM Sequence
- TGG
- Target locus
-
Chr.19: 6309758 - 6309780 on GRCm38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
89.96/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Cdc42bpg
- Entrez Gene ID
-
240505
- Gene Name
- CDC42 binding protein kinase gamma (DMPK-like)
- Gene Aliases
-
BC046418, MRCKgamma
- Chromosome Location
-
Chr.19: 6306457 - 6325655 on GRCm38
- UniGene ID
- Mm.329622
- Species
-
Mouse