gRNA Design Details
- Target DNA Sequence
- GTCATGTAGCATGTCGGCAT
- PAM Sequence
- AGG
- Target locus
-
Chr.8: 106217249 - 106217271 on GRCm38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
85.86/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Prmt7
- Entrez Gene ID
-
214572
- Gene Name
- protein arginine N-methyltransferase 7
- Gene Aliases
-
4933402B05Rik, BC006705
- Chromosome Location
-
Chr.8: 106211054 - 106251694 on GRCm38
- UniGene ID
- Mm.251804
- Species
-
Mouse