gRNA Design Details
- Target DNA Sequence
- TCGCCGACACCCAAAAAGCC
- PAM Sequence
- AGG
- Target locus
-
Chr.11: 59375519 - 59375541 on GRCm38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
88.03/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Prss38
- Entrez Gene ID
-
216797
- Gene Name
- protease, serine 38
- Gene Aliases
-
Gm249, Mpn2
- Chromosome Location
-
Chr.11: 59372669 - 59376253 on GRCm38
- UniGene ID
- Mm.339586
- Species
-
Mouse