gRNA Design Details

Target DNA Sequence
GCTGGCGCCCAAAATAAAGA
PAM Sequence
CGG
Target locus
Chr.11: 20364296 - 20364318 on GRCh38
Strand
Reverse
Application
Gene Knockout
Score: 69.88/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
HTATIP2
Entrez Gene ID
10553
Gene Name
HIV-1 Tat interactive protein 2
Gene Aliases
CC3, SDR44U1, TIP30
Chromosome Location
Chr.11: 20363685 - 20383783 on GRCh38
UniGene ID
Hs.90753
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001098520.1 NP_001091990.1 2
NM_001098521.1 NP_001091991.1 2
NM_001098522.1 NP_001091992.1 1
NM_001098523.1 NP_001091993.1 1
NM_006410.4 NP_006401.3 2