gRNA Design Details
- Target DNA Sequence
- GTAAGGGGCATTCCTGTTAC
- PAM Sequence
- AGG
- Target locus
-
Chr.4: 86856240 - 86856262 on GRCm38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
64.74/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Rps6
- Entrez Gene ID
-
20104
- Gene Name
- ribosomal protein S6
- Gene Aliases
-
S6R
- Chromosome Location
-
Chr.4: 86854099 - 86857367 on GRCm38
- UniGene ID
- Mm.379007
- Species
-
Mouse