gRNA Design Details
- Target DNA Sequence
- TGAGCGGACGGTTCAAAGAC
- PAM Sequence
- CGG
- Target locus
-
Chr.9: 45044187 - 45044209 on GRCm38
- Strand
- Forward
- Application
- Gene Knockout
Score:
87.28/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Mpzl2
- Entrez Gene ID
-
14012
- Gene Name
- myelin protein zero-like 2
- Gene Aliases
-
Eva, Eva1
- Chromosome Location
-
Chr.9: 45040291 - 45054043 on GRCm38
- UniGene ID
- Mm.33240
- Species
-
Mouse