gRNA Design Details
- Target DNA Sequence
- GGGCTATGCCGGATACTTGG
- PAM Sequence
- TGG
- Target locus
-
Chr.14: 69190942 - 69190964 on GRCm38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
88.62/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Nkx3-1
- Entrez Gene ID
-
18095
- Gene Name
- NK-3 transcription factor, locus 1 (Drosophila)
- Gene Aliases
-
Bax, NKX3.1, NKX3A, Nkx-3.1, bagpipe
- Chromosome Location
-
Chr.14: 69190692 - 69194658 on GRCm38
- UniGene ID
- Mm.3520
- Species
-
Mouse