gRNA Design Details
- Target DNA Sequence
- TGCCACATTAAAGACAAAGA
- PAM Sequence
- AGG
- Target locus
-
Chr.3: 7604074 - 7604096 on GRCm38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
75.74/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Il7
- Entrez Gene ID
-
16196
- Gene Name
- interleukin 7
- Gene Aliases
-
A630026I06Rik, Il-7, hlb368
- Chromosome Location
-
Chr.3: 7572028 - 7613427 on GRCm38
- UniGene ID
- Mm.3825
- Species
-
Mouse