gRNA Design Details
- Target DNA Sequence
- GCCGACGAGCGGAAAGATGA
- PAM Sequence
- AGG
- Target locus
-
Chr.1: 66380515 - 66380537 on GRCm38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
88.53/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Map2
- Entrez Gene ID
-
17756
- Gene Name
- microtubule-associated protein 2
- Gene Aliases
-
G1-397-34, MAP-2, Mtap-2, Mtap2, repro4
- Chromosome Location
-
Chr.1: 66175246 - 66442583 on GRCm38
- UniGene ID
- Mm.256966
- Species
-
Mouse