gRNA Design Details
- Target DNA Sequence
- GGAGATGATGACGATCTGCG
- PAM Sequence
- AGG
- Target locus
-
Chr.20: 33686154 - 33686176 on GRCh38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
70.59/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- E2F1
- Entrez Gene ID
-
1869
- Gene Name
- E2F transcription factor 1
- Gene Aliases
-
E2F-1, RBAP1, RBBP3, RBP3
- Chromosome Location
-
Chr.20: 33675486 - 33686404 on GRCh38
- UniGene ID
- Hs.654393
- Species
-
Human