gRNA Design Details
- Target DNA Sequence
- GTATGGCTCCTTTGCGCGCT
- PAM Sequence
- TGG
- Target locus
-
Chr.2: 119237832 - 119237854 on GRCm38
- Strand
- Forward
- Application
- Gene Knockout
Score:
86.66/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Spint1
- Entrez Gene ID
-
20732
- Gene Name
- serine protease inhibitor, Kunitz type 1
- Gene Aliases
-
HAI-1
- Chromosome Location
-
Chr.2: 119237360 - 119249535 on GRCm38
- UniGene ID
- Mm.104955
- Species
-
Mouse