gRNA Design Details
- Target DNA Sequence
- GGCTTCGGTGTCCAGCACAA
- PAM Sequence
- AGG
- Target locus
-
Chr.2: 119237947 - 119237969 on GRCm38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
85.33/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Spint1
- Entrez Gene ID
-
20732
- Gene Name
- serine protease inhibitor, Kunitz type 1
- Gene Aliases
-
HAI-1
- Chromosome Location
-
Chr.2: 119237360 - 119249535 on GRCm38
- UniGene ID
- Mm.104955
- Species
-
Mouse