gRNA Design Details
- Target DNA Sequence
- GATCTCCGCGATCTTCGCTA
- PAM Sequence
- AGG
- Target locus
-
Chr.11: 3266278 - 3266300 on GRCm38
- Strand
- Forward
- Application
- Gene Knockout
Score:
85.35/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Drg1
- Entrez Gene ID
-
13494
- Gene Name
- developmentally regulated GTP binding protein 1
- Gene Aliases
-
AA408859, AI132520, Nedd3
- Chromosome Location
-
Chr.11: 3249922 - 3266386 on GRCm38
- UniGene ID
- Mm.3250
- Species
-
Mouse