gRNA Design Details

Target DNA Sequence
CAAACATGTCCATGAGGGTC
PAM Sequence
CGG
Target locus
Chr.11: 27658184 - 27658206 on GRCh38
Strand
Reverse
Application
Gene Knockout
Score: 86.53/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
BDNF
Entrez Gene ID
627
Gene Name
brain derived neurotrophic factor
Gene Aliases
ANON2, BULN2
Chromosome Location
Chr.11: 27654893 - 27722058 on GRCh38
UniGene ID
Hs.502182
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001143805.1 NP_001137277.1 2
NM_001143806.1 NP_001137278.1 2
NM_001143807.1 NP_001137279.1 2
NM_001143808.1 NP_001137280.1 2
NM_001143809.1 NP_001137281.1 2
NM_001143810.1 NP_001137282.1 3
NM_001143811.1 NP_001137283.1 4
NM_001143812.1 NP_001137284.1 2
NM_001143813.1 NP_001137285.1 2
NM_001143814.1 NP_001137286.1 3
NM_001143816.1 NP_001137288.1 2
NM_001709.4 NP_001700.2 2
NM_170731.4 NP_733927.1 2
NM_170732.4 NP_733928.1 2
NM_170733.3 NP_733929.1 2
NM_170734.3 NP_733930.1 2
NM_170735.5 NP_733931.1 1
XM_011520280.2 XP_011518582.1 2