gRNA Design Details

Target DNA Sequence
GCACGAAGAACTCGAAGAGA
PAM Sequence
CGG
Target locus
Chr.19: 1020780 - 1020802 on GRCh38
Strand
Forward
Application
Gene Knockout
Score: 93.41/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
TMEM259
Entrez Gene ID
91304
Gene Name
transmembrane protein 259
Gene Aliases
ASBABP1, C19orf6, MBRL, MEMBRALIN, R32184_3
Chromosome Location
Chr.19: 1009651 - 1021169 on GRCh38
UniGene ID
Hs.515003
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001033026.1 NP_001028198.1 1
NM_033420.3 NP_219488.1 1
XM_005259675.2 XP_005259732.1 1
XM_005259676.2 XP_005259733.1 1
XM_005259677.2 XP_005259734.1 1
XM_005259678.2 XP_005259735.1 1
XM_017027457.1 XP_016882946.1 1