gRNA Design Details

Target DNA Sequence
AGCACGAGCAGAAGTTCACG
PAM Sequence
GGG
Target locus
Chr.19: 39169633 - 39169655 on GRCh38
Strand
Forward
Application
Gene Knockout
Score: 88.38/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
PAK4
Entrez Gene ID
10298
Gene Name
p21 (RAC1) activated kinase 4
Gene Aliases
-
Chromosome Location
Chr.19: 39125780 - 39179406 on GRCh38
UniGene ID
Hs.20447
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001014831.2 NP_001014831.1 4
NM_001014832.1 NP_001014832.1 2
NM_001014834.2 NP_001014834.1 2
NM_001014835.1 NP_001014835.1 3
NM_005884.3 NP_005875.1 3
XM_011526316.1 XP_011524618.1 3
XM_011526317.2 XP_011524619.1 3
XM_011526318.2 XP_011524620.1 3
XM_011526319.2 XP_011524621.1 3
XM_011526320.1 XP_011524622.1 3