Targeted Isoforms | Translated Protein | Targeted Exon(s) | |
---|---|---|---|
RefSeq | NM_001291428.1 | NP_001278357.1 | 4 |
NM_001291429.1 | NP_001278358.1 | 3 | |
NM_001291430.1 | NP_001278359.1 | 2 | |
NM_001291431.1 | NP_001278360.1 | 2 | |
NM_004324.3 | NP_004315.1 | 4 | |
NM_138761.3 | NP_620116.1 | 4 | |
NM_138763.3 | NP_620118.1 | 3 | |
NM_138764.4 | NP_620119.2 | 4 | |
NR_027882.1 | 4 | ||
XM_017027077.1 | XP_016882566.1 | 4 |