gRNA Design Details

Target DNA Sequence
GTTCAGTCTGCCACAGAACA
PAM Sequence
TGG
Target locus
Chr.21: 26112078 - 26112100 on GRCh38
Strand
Forward
Application
Gene Knockout
Score: 89.1/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
APP
Entrez Gene ID
351
Gene Name
amyloid beta precursor protein
Gene Aliases
AAA, ABETA, ABPP, AD1, APPI, CTFgamma, CVAP, PN-II, PN2
Chromosome Location
Chr.21: 25880550 - 26171128 on GRCh38
UniGene ID
Hs.434980
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_000484.3 NP_000475.1 2
NM_001136016.3 NP_001129488.1 2
NM_001136131.2 NP_001129603.1 2
NM_001204301.1 NP_001191230.1 2
NM_001204302.1 NP_001191231.1 2
NM_001204303.1 NP_001191232.1 2
NM_201413.2 NP_958816.1 2
NM_201414.2 NP_958817.1 2