gRNA Design Details

Target DNA Sequence
CTTCAAATCGTACAAACTGG
PAM Sequence
AGG
Target locus
Chr.6: 63285643 - 63285665 on GRCh38
Strand
Forward
Application
Gene Knock-Out
Score: 89.38/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
LGSN
Entrez Gene ID
51557
Gene Name
lengsin, lens protein with glutamine synthetase domain
Gene Aliases
GLULD1, LGS
Chromosome Location
Chr.6: 63275951 - 63567921 on GRCh38
UniGene ID
Hs.149585
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001143940.1 NP_001137412.1 3
NM_016571.2 NP_057655.2 3
XM_011535889.2 XP_011534191.1 9
XM_011535892.2 XP_011534194.1 9
XM_017010929.1 XP_016866418.1 10
XM_017010930.1 XP_016866419.1 9
XM_017010931.1 XP_016866420.1 8