gRNA Design Details
- Target DNA Sequence
- ATCTCTCGGATTGACCCAGA
- PAM Sequence
- TGG
- Target locus
-
Chr.3: 129739981 - 129740003 on GRCm38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
83.18/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Egf
- Entrez Gene ID
-
13645
- Gene Name
- epidermal growth factor
- Gene Aliases
-
AI790464
- Chromosome Location
-
Chr.3: 129677569 - 129755322 on GRCm38
- UniGene ID
- Mm.252481
- Species
-
Mouse