gRNA Design Details

Target DNA Sequence
ATAGACCTTGGAGCTGCTGT
PAM Sequence
GGG
Target locus
Chr.7: 36396326 - 36396348 on GRCh38
Strand
Reverse
Application
Gene Knock-Out
Score: 86.73/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
ANLN
Entrez Gene ID
54443
Gene Name
anillin actin binding protein
Gene Aliases
FSGS8, Scraps, scra
Chromosome Location
Chr.7: 36389803 - 36453791 on GRCh38
UniGene ID
Hs.62180
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001284301.2 NP_001271230.1 2
NM_001284302.2 NP_001271231.1 2
NM_018685.4 NP_061155.2 2
XM_006715746.1 XP_006715809.1 2
XM_006715747.3 XP_006715810.1 2
XM_017012354.1 XP_016867843.1 2
XM_017012355.1 XP_016867844.1 2
XM_017012356.1 XP_016867845.1 2