gRNA Design Details

Target DNA Sequence
CAGTGATTCTGTTGCTGTCC
PAM Sequence
CGG
Target locus
Chr.7: 36399245 - 36399267 on GRCh38
Strand
Forward
Application
Gene Knockout
Score: 85.76/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
ANLN
Entrez Gene ID
54443
Gene Name
anillin actin binding protein
Gene Aliases
FSGS8, Scraps, scra
Chromosome Location
Chr.7: 36389803 - 36453791 on GRCh38
UniGene ID
Hs.62180
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001284301.2 NP_001271230.1 3
NM_001284302.2 NP_001271231.1 3
NM_018685.4 NP_061155.2 3
XM_006715746.1 XP_006715809.1 3
XM_006715747.3 XP_006715810.1 3
XM_017012354.1 XP_016867843.1 3
XM_017012355.1 XP_016867844.1 3
XM_017012356.1 XP_016867845.1 3