gRNA Design Details
- Target DNA Sequence
- CAAGCAGTGTCTCGATGAGA
- PAM Sequence
- AGG
- Target locus
-
Chr.1: 1326961 - 1326983 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
91.24/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- CPTP
- Entrez Gene ID
-
80772
- Gene Name
- ceramide-1-phosphate transfer protein
- Gene Aliases
-
GLTPD1
- Chromosome Location
-
Chr.1: 1324756 - 1328896 on GRCh38
- UniGene ID
- Hs.515689
- Species
-
Human