gRNA Design Details

Target DNA Sequence
CAAGCAGTGTCTCGATGAGA
PAM Sequence
AGG
Target locus
Chr.1: 1326961 - 1326983 on GRCh38
Strand
Forward
Application
Gene Knockout
Score: 91.24/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
CPTP
Entrez Gene ID
80772
Gene Name
ceramide-1-phosphate transfer protein
Gene Aliases
GLTPD1
Chromosome Location
Chr.1: 1324756 - 1328896 on GRCh38
UniGene ID
Hs.515689
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001029885.1 NP_001025056.1 2
XM_005244801.3 XP_005244858.1 2
XM_011542200.2 XP_011540502.1 2