gRNA Design Details
- Target DNA Sequence
- CTCCATCGAGAACTCCGAAG
- PAM Sequence
- AGG
- Target locus
-
Chr.16: 57651303 - 57651325 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
89.81/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- ADGRG1
- Entrez Gene ID
-
9289
- Gene Name
- adhesion G protein-coupled receptor G1
- Gene Aliases
-
BFPP, BPPR, GPR56, TM7LN4, TM7XN1
- Chromosome Location
-
Chr.16: 57619535 - 57665039 on GRCh38
- UniGene ID
- Hs.513633
- Species
-
Human