gRNA Design Details
- Target DNA Sequence
- TGAGTTGGAGGTGTTGTACG
- PAM Sequence
- CGG
- Target locus
-
Chr.2: 203391079 - 203391101 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
87.67/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- ABI2
- Entrez Gene ID
-
10152
- Gene Name
- abl-interactor 2
- Gene Aliases
-
ABI-2, ABI2B, AIP-1, AblBP3, SSH3BP2, argBP1, argBPIA, argBPIB
- Chromosome Location
-
Chr.2: 203327874 - 203432173 on GRCh38
- UniGene ID
- Hs.471156
- Species
-
Human