gRNA Design Details

Target DNA Sequence
TGAGTTGGAGGTGTTGTACG
PAM Sequence
CGG
Target locus
Chr.2: 203391079 - 203391101 on GRCh38
Strand
Reverse
Application
Gene Knock-Out
Score: 87.67/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
ABI2
Entrez Gene ID
10152
Gene Name
abl-interactor 2
Gene Aliases
ABI-2, ABI2B, AIP-1, AblBP3, SSH3BP2, argBP1, argBPIA, argBPIB
Chromosome Location
Chr.2: 203327874 - 203432173 on GRCh38
UniGene ID
Hs.471156
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001282925.1 NP_001269854.1 5
NM_001282926.1 NP_001269855.1 4
NM_001282927.1 NP_001269856.1 4
NM_001282932.1 NP_001269861.1 5
NM_005759.5 NP_005750.4 4
XM_005246217.2 XP_005246274.1 5
XM_005246218.2 XP_005246275.1 4
XM_005246221.2 XP_005246278.1 4
XM_005246225.2 XP_005246282.1 4
XM_005246226.2 XP_005246283.1 4
XM_006712174.2 XP_006712237.1 5
XM_006712177.2 XP_006712240.1 6
XM_006712179.2 XP_006712242.1 5
XM_006712180.2 XP_006712243.1 5
XM_006712181.2 XP_006712244.1 5
XM_006712182.2 XP_006712245.1 5
XM_006712185.1 XP_006712248.1 5
XM_006712186.2 XP_006712249.1 4
XM_011510472.2 XP_011508774.1 5
XM_011510473.2 XP_011508775.1 4
XM_017003118.1 XP_016858607.1 5
XM_017003119.1 XP_016858608.1 5
XM_017003120.1 XP_016858609.1 5
XM_017003121.1 XP_016858610.1 5
XM_017003122.1 XP_016858611.1 5
XM_017003123.1 XP_016858612.1 4
XM_017003124.1 XP_016858613.1 4
XM_017003125.1 XP_016858614.1 4
XM_017003126.1 XP_016858615.1 4
XM_017003127.1 XP_016858616.1 4
XM_017003128.1 XP_016858617.1 6
XM_017003129.1 XP_016858618.1 5
XM_017003130.1 XP_016858619.1 5
XM_017003131.1 XP_016858620.1 4
XM_017003132.1 XP_016858621.1 5
XM_017003133.1 XP_016858622.1 6
XM_017003134.1 XP_016858623.1 4
XM_017003135.1 XP_016858624.1 4
XM_017003136.1 XP_016858625.1 4
XM_017003137.1 XP_016858626.1 6
XM_017003138.1 XP_016858627.1 5
XM_017003139.1 XP_016858628.1 5
XM_017003140.1 XP_016858629.1 5
XM_017003141.1 XP_016858630.1 4
XM_017003142.1 XP_016858631.1 4
XM_017003143.1 XP_016858632.1 6
XM_017003144.1 XP_016858633.1 5
XM_017003145.1 XP_016858634.1 4
XM_017003146.1 XP_016858635.1 4