gRNA Design Details

Target DNA Sequence
GTTCGTTATGAGAAGAGATA
PAM Sequence
TGG
Target locus
Chr.4: 85942095 - 85942117 on GRCh38
Strand
Forward
Application
Gene Knockout
Score: 87.67/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
ARHGAP24
Entrez Gene ID
83478
Gene Name
Rho GTPase activating protein 24
Gene Aliases
FILGAP, RC-GAP72, RCGAP72, p73, p73RhoGAP
Chromosome Location
Chr.4: 85475131 - 86002670 on GRCh38
UniGene ID
Hs.444229
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001025616.2 NP_001020787.2 5
NM_001042669.1 NP_001036134.1 3
NM_001287805.1 NP_001274734.1 3
NM_031305.2 NP_112595.2 2
XM_011532300.2 XP_011530602.1 3