gRNA Design Details
- Target DNA Sequence
- GAATATGCCTACGGTTCAGC
- PAM Sequence
- AGG
- Target locus
-
Chr.12: 2797806 - 2797828 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
87.26/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- FKBP4
- Entrez Gene ID
-
2288
- Gene Name
- FK506 binding protein 4
- Gene Aliases
-
FKBP51, FKBP52, FKBP59, HBI, Hsp56, PPIase, p52
- Chromosome Location
-
Chr.12: 2794942 - 2805423 on GRCh38
- UniGene ID
- Hs.524183
- Species
-
Human