gRNA Design Details
- Target DNA Sequence
- CTGATGTCCACTCCCTCCAT
- PAM Sequence
- GGG
- Target locus
-
Chr.12: 2795194 - 2795216 on GRCh38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
83.78/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- FKBP4
- Entrez Gene ID
-
2288
- Gene Name
- FK506 binding protein 4
- Gene Aliases
-
FKBP51, FKBP52, FKBP59, HBI, Hsp56, PPIase, p52
- Chromosome Location
-
Chr.12: 2794942 - 2805423 on GRCh38
- UniGene ID
- Hs.524183
- Species
-
Human